Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134156.2 | NP_598917.1 | 16, 17 | 2240 | ||
GenBank mRNA | AK142591.1 | - | 7, 8 | 956 | ||
AK154281.1 | - | 16, 17 | 2233 | |||
BC003232.1 | - | 4, 5 | 485 | |||
BC023206.1 | - | 3, 4 | 442 | |||
BC054830.1 | - | 16, 17 | 2185 | 6308412 | ||
DQ288940.1 | - | 16, 17 | 1960 |