Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Braf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139294.5 | NP_647455.3 | 7, 8 | 989 | ||
GenBank mRNA | AJ276307.1 | - | 5, 6 | 673 | ||
AK140431.1 | - | 7, 8 | 990 |