Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145588.1 | NP_663563.1 | 12 | 1731 | ||
GenBank mRNA | AK075922.1 | - | 12 | 1722 | ||
AK084310.1 | - | 12 | 1720 | |||
AK088033.1 | - | 9 | 1307 | |||
AK205411.1 | - | 3 | 181 | |||
BC003427.1 | - | 12 | 1731 | 2615715 |