Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153781.1 | NP_722476.1 | 14 | 1824 | ||
GenBank mRNA | AK032769.1 | - | 14 | 1837 | ||
AK035673.1 | - | 14 | 1554 | |||
AK133794.1 | - | 5 | 554 | |||
AK134667.1 | - | 14 | 1834 | |||
AK142716.1 | - | 14 | 1834 | |||
AK147763.1 | - | 14 | 1856 | |||
AK168381.1 | - | 14 | 1836 | |||
AK169052.1 | - | 14 | 1859 | |||
AK169631.1 | - | 14 | 1838 | |||
BC031457.1 | - | 14 | 1831 | |||
BC032209.1 | - | 8 | 979 | |||
BC035283.1 | - | 14 | 1824 | 4947455 |