Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153781.1 | NP_722476.1 | 14 | 1780 | ||
GenBank mRNA | AK032769.1 | - | 14 | 1793 | ||
AK035673.1 | - | 14 | 1510 | |||
AK133794.1 | - | 5 | 510 | |||
AK134667.1 | - | 14 | 1790 | |||
AK142716.1 | - | 14 | 1790 | |||
AK147763.1 | - | 14 | 1812 | |||
AK168381.1 | - | 14 | 1792 | |||
AK169052.1 | - | 14 | 1815 | |||
AK169631.1 | - | 14 | 1794 | |||
BC031457.1 | - | 14 | 1787 | |||
BC032209.1 | - | 8 | 935 | |||
BC035283.1 | - | 14 | 1780 | 4947455 |