Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133198.2 | NP_573461.2 | 11 | 1382 | ||
GenBank mRNA | AF288783.1 | - | 11 | 1324 | ||
AK140321.1 | - | 11 | 1329 | |||
AK145989.1 | - | 11 | 1383 | |||
AK149491.1 | - | 1557 | ||||
AK167337.1 | - | 11 | 1329 | |||
BC013636.1 | - | 11 | 1316 | 4196237 |