Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286573.1 | NP_001273502.1 | 19 | 2760 | ||
NM_001286575.1 | NP_001273504.1 | 21 | 2698 | |||
NM_145830.2 | NP_665829.1 | 20 | 2862 | |||
NM_147151.2 | NP_671493.1 | 20 | 2596 | |||
GenBank mRNA | AB077209.1 | - | 20 | 2589 | ||
AB077210.1 | - | 20 | 2862 | |||
AK077743.1 | - | 8 | 918 | |||
AK154246.1 | - | 19 | 2783 | |||
AK182117.1 | - | 3 | 285 | |||
BC051494.1 | - | 1 | 122 | |||
BC058357.1 | - | 21 | 2696 | |||
BC096670.1 | - | 6 | 656 |