Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253712.1 | NP_001240641.1 | 3 | 438 | ||
NM_001253714.1 | NP_001240643.1 | 2 | 355 | |||
NM_001253716.1 | NP_001240645.1 | 3 | 166 | |||
NM_026322.4 | NP_080598.2 | 8 | 837 | |||
GenBank mRNA | AK002356.1 | - | 3 | 313 | ||
AK009822.1 | - | 3 | 315 | |||
AK018338.1 | - | 3 | 160 | |||
AK030278.1 | - | 3 | 316 | |||
AK049714.1 | - | 3 | 439 | |||
BB652761.1 | - | 3 | 436 | |||
BC089311.1 | - | 8 | 837 | 6412625 | ||
BY282106.1 | - | 3 | 173 | |||
CB953198.1 | - | 2 | 226 |