Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync2h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029851.2 | NP_084127.2 | 26 | 3917 | ||
GenBank mRNA | AY452064.1 | - | 4209 | |||
DQ104402.1 | - | 26 | 3917 |