Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290684.1 | NP_001277613.1 | 7 | 3270 | ||
NM_001290685.1 | NP_001277614.1 | 3 | 595 | |||
NM_001290686.1 | NP_001277615.1 | 3 | 402 | |||
NM_001290687.1 | NP_001277616.1 | 3 | 584 | |||
NM_172441.3 | NP_766029.2 | 7 | 3288 | |||
GenBank mRNA | AK029338.1 | - | 3 | 276 | ||
AK032256.1 | - | 3 | 277 | |||
AK049850.1 | - | 3 | 334 | |||
BC092373.1 | - | 3 | 590 | |||
CF534250.1 | - | 3 | 545 | |||
CK635066.1 | - | 4 | 553 | |||
CO431718.1 | - | 3 | 584 | |||
DQ358971.1 | - | 7 | 3251 | |||
EF071946.1 | - | 7 | 3288 |