Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133882.2 | NP_598643.1 | 7 | 1080 | ||
GenBank mRNA | AB077304.1 | - | 7 | 1002 | ||
AB077305.1 | - | 7 | 1002 | |||
AB077306.1 | - | 6 | 804 | |||
AK050313.1 | - | 7 | 1081 | |||
BC022129.1 | - | 5 | 711 | |||
BC096382.1 | - | 7 | 1054 | 5102825 |