Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adarb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024837.2 | NP_001020008.1 | 4 | 1141 | ||
NM_130895.3 | NP_570965.2 | 4 | 1141 | |||
GenBank mRNA | AF291049.1 | - | 3 | 976 | ||
AF403106.2 | - | 4 | 1093 | |||
AF403107.2 | - | 4 | 1093 | |||
AF403108.2 | - | 4 | 1140 | |||
AF403109.2 | - | 4 | 1140 | |||
AF525421.1 | - | 5 | 1166 | |||
AK138318.1 | - | 4 | 1134 | |||
AK141777.1 | - | 4 | 1113 | |||
AK147298.1 | - | 5 | 1220 | |||
AY162454.1 | - | 5 | 1166 | |||
BC051107.1 | - | 4 | 1126 |