Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012026.2 | NP_036156.2 | 31 | 4143 | ||
GenBank mRNA | AK029167.1 | - | 25 | 3303 | ||
AK034079.1 | - | 1 | 2912 | |||
BC098331.1 | - | 31 | 4132 | |||
BC138030.1 | - | 31 | 4067 | 8861049 | ||
U73199.1 | - | 31 | 4143 |