Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cse1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023565.3 | NP_076054.1 | 17, 18 | 1911 | ||
GenBank mRNA | AF301152.1 | - | 17, 18 | 1911 | ||
AK138940.1 | - | 3, 4 | 335 | |||
BC145393.1 | - | 16, 17 | 1682 |