Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cse1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023565.3 | NP_076054.1 | 19 | 2136 | ||
GenBank mRNA | AF301152.1 | - | 19 | 2136 | ||
AK138940.1 | - | 5 | 560 | |||
BC145393.1 | - | 18 | 1907 |