Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tshz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081300.1 | NP_001074769.1 | 2 | 2820 | ||
GenBank mRNA | AF191309.1 | - | 1 | 1619 | ||
AK220545.1 | - | 2 | 2662 | |||
BC017636.1 | - | 1 | 671 | |||
BC038610.1 | - | 1 | 827 | |||
BC040215.1 | - | 2 | 2009 | |||
BC049795.1 | - | 1 | 1260 | |||
BC049799.1 | - | 1 | 1260 | |||
BC058264.1 | - | 1 | 821 | 6488400 |