Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tshz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081300.1 | NP_001074769.1 | 2 | 4351 | ||
GenBank mRNA | AF191309.1 | - | 1 | 3150 | ||
AK017514.1 | - | 1 | 961 | |||
AK220545.1 | - | 2 | 4193 | |||
BC017636.1 | - | 1 | 2202 | |||
BC038610.1 | - | 1 | 2358 | |||
BC040215.1 | - | 2 | 3540 | |||
BC049795.1 | - | 1 | 2791 | |||
BC049799.1 | - | 1 | 2791 | |||
BC058264.1 | - | 1 | 2352 | 6488400 |