Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145129.2 | NP_660111.2 | 5 | 1256 | ||
GenBank mRNA | AF459029.1 | - | 5 | 1229 | ||
AF472588.1 | - | 5 | 1158 | |||
AK041217.1 | - | 735 | ||||
AK051730.1 | - | 5 | 1260 | |||
AK053497.1 | - | 5 | 1184 | |||
AK080415.1 | - | 5 | 1257 | |||
AK080475.1 | - | 5 | 1260 | |||
AK148786.1 | - | 5 | 1228 | |||
AY574262.1 | - | 5 | 1091 | |||
BC120597.1 | - | 5 | 1127 | 40129520 | ||
BC120599.1 | - | 5 | 1127 | 40129522 |