Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138748.5 | NP_620087.3 | 2 | 311 | ||
GenBank mRNA | AK031591.1 | - | 2 | 312 | ||
AK046288.1 | - | 2 | 115 | |||
AK049184.1 | - | 2 | 302 | |||
AK150029.1 | - | 2 | 306 | |||
AK160980.1 | - | 2 | 166 | |||
AK179985.1 | - | 1 | 34 | |||
AK207653.1 | - | 1 | 34 | |||
AK212725.1 | - | 1 | 34 | |||
AK215573.1 | - | 1 | 34 | |||
AK219416.1 | - | 1 | 34 | |||
AK219614.1 | - | 1 | 34 | |||
AY035997.1 | - | 2 | 164 | |||
BC006626.1 | - | 2 | 227 | 3158086 | ||
BC052852.1 | - | 2 | 237 | 30058414 |