Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153054.2 | NP_694694.1 | 5 | 971 | ||
GenBank mRNA | AK049514.1 | - | 3 | 857 | ||
AK076433.1 | - | 4 | 695 | |||
AK144034.1 | - | 4 | 695 | |||
AY779336.1 | - | 3 | 629 | |||
BC027192.1 | - | 5 | 909 | 4239930 | ||
BC031146.1 | - | 5 | 939 | 4974776 |