Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139145.4 | NP_631884.1 | 6 | 1642 | ||
GenBank mRNA | AB066227.1 | - | 5 | 1502 | ||
AK052424.1 | - | 7 | 1938 | |||
AK158212.1 | - | 7 | 1759 | |||
AK168925.1 | - | 3 | 537 | |||
BC050090.1 | - | 6 | 1643 | 30012868 |