Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033074.3 | NP_149065.2 | 14, 15 | 1747 | ||
GenBank mRNA | AK010256.1 | - | 14, 15 | 1714 | ||
AK011146.2 | - | 14, 15 | 1748 | |||
AK031064.1 | - | 14, 15 | 1711 | |||
AK152945.1 | - | 14, 15 | 1708 | |||
AK153314.1 | - | 14, 15 | 1719 | |||
AK166693.1 | - | 14, 15 | 1714 | |||
AK167597.1 | - | 14, 15 | 1756 | |||
AK168969.1 | - | 14, 15 | 1707 | |||
BC004621.1 | - | 7, 8 | 834 | |||
BC055371.1 | - | 14, 15 | 1733 | 30060749 | ||
BC060176.1 | - | 1842 |