Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009592.1 | NP_033722.1 | 12, 13 | 1686 | ||
GenBank mRNA | AK040069.1 | - | 12, 13 | 1683 | ||
AK084853.1 | - | 12, 13 | 1677 | |||
AK151967.1 | - | 12, 13 | 1672 | |||
AK152535.1 | - | 12, 13 | 1672 | |||
AK152816.1 | - | 12, 13 | 1672 | |||
AK152946.1 | - | 12, 13 | 1674 | |||
BC035534.1 | - | 9, 10 | 1315 | |||
BC151037.1 | - | 12, 13 | 1686 | 9087950 | ||
U43892.1 | - | 11, 12 | 1490 |