Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Laptm4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033521.3 | NP_277056.1 | 1, 2 | 391 | ||
GenBank mRNA | AF317418.1 | - | 1, 2 | 93 | ||
AK077269.1 | - | 1, 2 | 254 | |||
AK077933.1 | - | 1, 2 | 391 | |||
AK079581.1 | - | 1, 2 | 254 | |||
AK082807.1 | - | 1, 2 | 270 | |||
AK166014.1 | - | 1, 2 | 293 | |||
BC019120.1 | - | 1, 2 | 261 | 4507607 | ||
EU561010.1 | - | 1, 2 | 391 |