Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286607.1 | NP_001273536.1 | 2 | 338 | ||
NM_053242.4 | NP_444472.2 | 4 | 675 | |||
NM_212435.1 | NP_997600.1 | 2 | 338 | |||
GenBank mRNA | AK051668.1 | - | 4 | 637 | ||
AK080549.1 | - | 4 | 546 | |||
AK082361.1 | - | 4 | 347 | |||
AK164319.1 | - | 2 | 325 | |||
AY079003.1 | - | 4 | 304 | |||
BC058960.1 | - | 2 | 335 | 6811877 | ||
BC062926.1 | - | 2 | 338 | 6851153 |