Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc33a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272035.1 | NP_001258964.1 | 1 | 746 | ||
NM_015728.5 | NP_056543.2 | 1 | 746 | |||
GenBank mRNA | AB016795.1 | - | 1 | 203 | ||
AK135092.1 | - | 1 | 675 | |||
AK136323.1 | - | 1 | 741 | |||
AK144703.1 | - | 1 | 612 | |||
BC005616.1 | - | 1 | 323 | 3154886 | ||
BC054395.1 | - | 1 | 746 | 6492236 | ||
BC063104.1 | - | 679 | 5042025 |
Gene Symbol: D630022N01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052681.1 | - | 1 | 353 | ||
AK085339.1 | - | 1 | 351 |