Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080633.2 | NP_542364.1 | 9, 10 | 1198 | ||
GenBank mRNA | AK143917.1 | - | 9, 10 | 1177 | ||
AK144207.1 | - | 9, 10 | 1161 | |||
AK145511.1 | - | 9, 10 | 1178 | |||
AK150027.1 | - | 9, 10 | 1178 | |||
AK150954.1 | - | 10, 11 | 1162 | |||
AK165411.1 | - | 9, 10 | 1162 | |||
AK191827.1 | - | 3, 4 | 264 | |||
AK198314.1 | - | 3, 4 | 244 | |||
AK198988.1 | - | 3, 4 | 268 | |||
AK199021.1 | - | 4, 5 | 339 | |||
AK206733.1 | - | 3, 4 | 254 | |||
BC004645.1 | - | 9, 10 | 1224 | 3158239 | ||
BC094462.1 | - | 9, 10 | 1157 | 4238599 | ||
CA750686.1 | - | 5, 6 | 584 | |||
CO426953.1 | - | 2, 3 | 132 |