Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102404.1 | NP_001095874.1 | 5 | 1040 | ||
NM_001102405.1 | NP_001095875.1 | 5 | 1045 | |||
NM_007388.3 | NP_031414.1 | 5 | 987 | |||
GenBank mRNA | AK008391.1 | - | 4 | 767 | ||
AK010865.1 | - | 5 | 1009 | |||
AK011915.1 | - | 5 | 1010 | |||
BC012911.1 | - | 5 | 939 | 3588062 | ||
BC019160.1 | - | 5 | 1003 | 3596147 | ||
BC029644.1 | - | 5 | 939 | 3584530 | ||
BQ031772.1 | - | 1 | 377 |