Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007389.5 | NP_031415.2 | 6 | 782 | ||
GenBank mRNA | AK029177.1 | - | 6 | 751 | ||
AK132589.1 | - | 6 | 722 | |||
BC125439.1 | - | 6 | 782 | 40129954 | ||
BC137678.1 | - | 6 | 782 | 8860697 | ||
X03986.1 | - | 6 | 739 |