Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015730.5 | NP_056545.3 | 5, 6 | 1858 | ||
GenBank mRNA | AF225912.2 | - | 5, 6 | 1756 | ||
AK034228.1 | - | 5, 6 | 1869 | |||
AK083157.1 | - | 5, 6 | 1789 | |||
AK136886.1 | - | 5, 6 | 1845 | |||
AK143938.1 | - | 5, 6 | 1866 | |||
AY574263.1 | - | 5, 6 | 1754 | |||
BC053013.1 | - | 5, 6 | 1858 | 5706867 |