Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053260.3 | NP_444490.2 | 3, 4 | 417 | ||
GenBank mRNA | AF305425.2 | - | 2, 3 | 253 | ||
AF442819.1 | - | 2, 3 | 253 | |||
AK144368.1 | - | 3, 4 | 418 | |||
BC104122.1 | - | 2, 3 | 253 | 40044207 | ||
BC104123.1 | - | 2, 3 | 253 |