Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053268.2 | NP_444498.2 | 6 | 553 | ||
GenBank mRNA | AB056433.1 | - | 6 | 546 | ||
AK034434.1 | - | 6 | 553 | |||
AK154499.1 | - | 6 | 570 | |||
AK166654.1 | - | 6 | 553 |