Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033523.4 | NP_277058.1 | 4, 5 | 937 | ||
GenBank mRNA | AB063496.1 | - | 4, 5 | 448 | ||
AK036430.1 | - | 4, 5 | 938 | |||
AK153700.1 | - | 4, 5 | 965 | |||
AK157817.1 | - | 4, 5 | 940 | |||
BC066013.1 | - | 4, 5 | 1007 | 5706237 |