Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007395.3 | NP_031421.1 | 5 | 888 | ||
GenBank mRNA | AK154578.1 | - | 911 | |||
AK157910.1 | - | 5 | 889 | |||
BC059832.1 | - | 881 | ||||
BC145775.1 | - | 5 | 941 | 40131078 | ||
BC145777.1 | - | 5 | 941 | 40131080 | ||
Z31663.1 | - | 5 | 877 |