Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007399.3 | NP_031425.2 | 14 | 2146 | ||
GenBank mRNA | AF011379.1 | - | 14 | 1967 | ||
AK036599.1 | - | 8 | 1190 | |||
AK036883.1 | - | 6 | 834 | |||
AK036918.1 | - | 6 | 835 | |||
BC057346.1 | - | 14 | 2107 |