Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272057.1 | NP_001258986.1 | 6 | 783 | ||
NM_001272058.1 | NP_001258987.1 | 6 | 783 | |||
NM_001272059.1 | NP_001258988.1 | 6 | 783 | |||
NM_007401.3 | NP_031427.2 | 6 | 783 | |||
GenBank mRNA | AK016574.1 | - | 6 | 439 | ||
AK029519.1 | - | 6 | 473 | |||
AK161262.1 | - | 6 | 783 | |||
AK161415.1 | - | 3 | 161 | |||
AK161417.1 | - | 6 | 453 | |||
BC094237.1 | - | 6 | 480 | 30604517 | ||
U22059.1 | - | 6 | 447 |