Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089086.1 | - | 2 | 94 | ||
AK089169.1 | - | 2 | 116 | |||
AK150411.1 | - | 2 | 96 | |||
AK152675.1 | - | 2 | 113 | |||
AK152769.1 | - | 2 | 95 | |||
AK153065.1 | - | 2 | 95 | |||
AK155772.1 | - | 2 | 94 | |||
AK155912.1 | - | 2 | 114 | |||
AK160041.1 | - | 2 | 97 | |||
AK170019.1 | - | 2 | 119 | |||
AK171022.1 | - | 2 | 116 | |||
AK171060.1 | - | 2 | 94 | |||
BC025584.1 | - | 2 | 148 | 5324943 | ||
X13335.1 | - | 2 | 93 |