Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089086.1 | - | 16, 17 | 1799 | ||
AK089169.1 | - | 16, 17 | 1839 | |||
AK134118.1 | - | 6, 7 | 1353 | |||
AK150411.1 | - | 16, 17 | 1801 | |||
AK151535.1 | - | 8, 9 | 1030 | |||
AK152675.1 | - | 16, 17 | 1818 | |||
AK152769.1 | - | 16, 17 | 1800 | |||
AK153065.1 | - | 16, 17 | 1800 | |||
AK155772.1 | - | 16, 17 | 1799 | |||
AK155912.1 | - | 16, 17 | 1819 | |||
AK160041.1 | - | 16, 17 | 1802 | |||
AK170019.1 | - | 16, 17 | 1824 | |||
AK171022.1 | - | 14, 15 | 2376 | |||
AK171060.1 | - | 16, 17 | 1799 | |||
BC025584.1 | - | 16, 17 | 1853 | 5324943 | ||
X13335.1 | - | 16, 17 | 1798 |