Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037723.3 | NP_001032812.2 | 5 | 815 | ||
NM_001037724.4 | NP_001032813.1 | 5 | 751 | |||
NM_001109756.1 | NP_001103226.1 | 5 | 719 | |||
NM_007406.2 | NP_031432.2 | 5 | 903 | |||
GenBank mRNA | AK154262.1 | - | 5 | 806 | ||
AK154652.1 | - | 5 | 721 | |||
AK155646.1 | - | 5 | 720 | |||
AK155831.1 | - | 5 | 722 | |||
BC115833.1 | - | 3 | 597 | 40086640 | ||
U12919.1 | - | 5 | 904 |