Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291903.1 | NP_001278832.1 | 1 | 1601 | ||
NM_009623.2 | NP_033753.2 | 1 | 1601 | |||
GenBank mRNA | AK138244.1 | - | 1 | 1602 | ||
AK163443.1 | - | 1 | 1577 | |||
BC059010.1 | - | 1 | 699 | |||
BC094231.1 | - | 1 | 1521 | |||
U85021.1 | - | 1 | 1461 |