Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243041.1 | NP_001229970.1 | 7 | 726 | ||
NM_134079.4 | NP_598840.1 | 7 | 683 | |||
GenBank mRNA | AK032871.1 | - | 7 | 641 | ||
AK151863.1 | - | 7 | 728 | |||
AK160264.1 | - | 7 | 680 | |||
AY540996.1 | - | 7 | 625 | |||
AY540997.1 | - | 6 | 570 | |||
BC009659.1 | - | 7 | 648 | 3485959 |