Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adssl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007421.2 | NP_031447.1 | 11 | 1180 | ||
GenBank mRNA | AK089520.1 | - | 11 | 1155 | ||
AK146907.1 | - | 11 | 1183 | |||
AK149634.1 | - | 11 | 1155 | |||
AK150874.1 | - | 11 | 1151 | |||
BC039943.1 | - | 11 | 1211 | 4014672 | ||
M74495.1 | - | 11 | 1138 |