Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF053943.1 | - | 18 | 2449 | ||
AK004712.1 | - | 14 | 1458 | |||
AK128980.1 | - | 18 | 2525 | |||
AK158276.1 | - | 18 | 2475 | |||
AK159330.1 | - | 18 | 2456 | |||
AK159342.1 | - | 18 | 2475 | |||
AK159377.1 | - | 18 | 2446 | |||
AK159409.1 | - | 18 | 2476 | |||
AK159774.1 | - | 18 | 2447 | |||
AK159957.1 | - | 18 | 2447 | |||
AK205582.1 | - | 1 | 42 | |||
BC082577.1 | - | 18 | 2461 | 30648530 | ||
X80478.1 | - | 9 | 1179 |