Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF053943.1 | - | 21 | 3416 | ||
AK004712.1 | - | 17 | 2425 | |||
AK128980.1 | - | 21 | 3492 | |||
AK158276.1 | - | 21 | 3442 | |||
AK159330.1 | - | 21 | 3423 | |||
AK159342.1 | - | 21 | 3442 | |||
AK159377.1 | - | 21 | 3413 | |||
AK159409.1 | - | 21 | 3443 | |||
AK159774.1 | - | 21 | 3414 | |||
AK159957.1 | - | 21 | 3414 | |||
AK207752.1 | - | 1 | 119 | |||
AK216208.1 | - | 2 | 295 | |||
BC082577.1 | - | 21 | 3428 | 30648530 | ||
X80478.1 | - | 12 | 2146 |