Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178803.2 | NP_848918.1 | 5, 6 | 1025 | ||
GenBank mRNA | AB221647.1 | - | 5, 6 | 676 | ||
AF090326.1 | - | 760 | ||||
AK029011.1 | - | 5, 6 | 1027 | |||
AK045838.1 | - | 5, 6 | 791 | |||
AK045987.1 | - | 5, 6 | 791 | |||
AK155272.1 | - | 5, 6 | 757 | |||
BC025467.1 | - | 5, 6 | 1483 | 5351959 | ||
BC031376.1 | - | 6, 7 | 886 | 5356616 | ||
BC057681.1 | - | 5, 6 | 805 | 3499299 |