Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007423.4 | NP_031449.3 | 6 | 711 | ||
GenBank mRNA | AK010934.1 | - | 6 | 687 | ||
AK075972.1 | - | 6 | 691 | |||
AK076053.1 | - | 6 | 687 | |||
AK076197.1 | - | 6 | 689 | |||
AK145975.1 | - | 6 | 687 | |||
AK146513.1 | - | 6 | 689 | |||
AK146524.1 | - | 6 | 689 | |||
AK167549.1 | - | 6 | 687 | |||
AK168426.1 | - | 6 | 690 | |||
AK168574.1 | - | 6 | 689 | |||
AK168576.1 | - | 6 | 687 | |||
AK168751.1 | - | 6 | 689 | |||
AK168818.1 | - | 5 | 565 | |||
AK168883.1 | - | 6 | 687 | |||
AK169046.1 | - | 6 | 689 | |||
AK169054.1 | - | 6 | 689 | |||
AK169147.1 | - | 6 | 687 | |||
BC066206.1 | - | 6 | 674 | 30469872 | ||
V00743.1 | - | 6 | 685 |