Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013464.4 | NP_038492.1 | 8, 9 | 1358 | ||
GenBank mRNA | AF405560.1 | - | 8, 9 | 992 | ||
AF405561.1 | - | 8, 9 | 992 | |||
AF405562.1 | - | 8, 9 | 992 | |||
AF405563.1 | - | 8, 9 | 992 | |||
AF405565.1 | - | 8, 9 | 992 | |||
AF405566.1 | - | 8, 9 | 992 | |||
AF405567.1 | - | 8, 9 | 992 | |||
AF405568.1 | - | 8, 9 | 992 | |||
AF405570.1 | - | 8, 9 | 992 | |||
AK136072.1 | - | 2, 3 | 242 | |||
AK153678.1 | - | 8, 9 | 1359 | |||
AK155380.1 | - | 8, 9 | 1410 | |||
AY662080.1 | - | 8, 9 | 992 | |||
AY662081.1 | - | 7, 8 | 888 | |||
AY662082.1 | - | 8, 9 | 992 | |||
AY662083.1 | - | 8, 9 | 992 | |||
D38416.1 | - | 8, 9 | 1007 |