Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009654.3 | NP_033784.2 | 9 | 1159 | ||
GenBank mRNA | AJ011413.1 | - | 9 | 1146 | ||
AJ457860.1 | - | 9 | 1113 | |||
AK010025.1 | - | 9 | 1148 | |||
AK050248.1 | - | 9 | 1148 | |||
AK050644.1 | - | 8 | 1017 | |||
AK131891.1 | - | 9 | 1151 | |||
AK160487.1 | - | 9 | 1151 | |||
BC024643.1 | - | 9 | 1133 | 4242076 | ||
BC049971.1 | - | 9 | 1133 | 4161986 | ||
M16111.1 | - | 6 | 819 |