Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009654.3 | NP_033784.2 | 7 | 837 | ||
GenBank mRNA | AJ011413.1 | - | 7 | 824 | ||
AJ457860.1 | - | 7 | 791 | |||
AK010025.1 | - | 7 | 826 | |||
AK050248.1 | - | 7 | 826 | |||
AK050644.1 | - | 6 | 695 | |||
AK131891.1 | - | 7 | 829 | |||
AK160487.1 | - | 7 | 829 | |||
BC024643.1 | - | 7 | 811 | 4242076 | ||
BC049971.1 | - | 7 | 811 | 4161986 | ||
M16111.1 | - | 4 | 497 |