Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009655.2 | NP_033785.1 | 3 | 735 | ||
GenBank mRNA | AB003026.1 | - | 3 | 397 | ||
AK030851.1 | - | 3 | 736 | |||
AK031391.1 | - | 1811 | ||||
AY644766.1 | - | 3 | 392 | |||
BC027280.1 | - | 3 | 392 | 3501215 | ||
U95030.1 | - | 3 | 203 |